
(S)-(-)-2-Amino-2'-hydroxy-1,1'-binaphthyl ≥98.0% (by HPLC)
Catalog # TCIAA2317-100MG
Supplier: TCI
CAS Number:
Specifications
- VPE:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:171.0 to 175.0 deg-C
Specification Test Results
About this item
:
:285.35 g/mol
:GHS05
CAS Number:137848-29-4
Formula:C₂₀H₁₅NO
MDL Number:MFCD01882346