
Artemether ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:71963-77-4
:
Formula:C₁₆H₂₆O₅
MDL Number:MFCD00866205
:298.38 g/mol
Merck Index:14,00815
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:86.0 to 90.0 deg-C
- Specific rotation [a]20/D:+166 to +173 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together