Order Entry
Netherlands
ContactUsLinkComponent
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
Catalog # TCIAA2121-5G
Supplier:  TCI
CAS Number:  
4-(Aminomethyl)cyclohexanecarboxylic acid (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
Catalog # TCIAA2121-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 % (total of isomer)
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :157.21 g/mol
Melting Pt:233°C
CAS Number:701-54-2
Formula:C₈H₁₅NO₂
MDL Number:MFCD00064951