2-Allenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:865350-17-0
:
Formula:C₉H₁₅BO₂
MDL Number:MFCD09265121
:166.03 g/mol
Boiling Pt:81°C
Flash Pt:66°C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together