
3'-Azido-3'-deoxythymidine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:30516-87-1
:
Formula:C₁₀H₁₃N₅O₄
MDL Number:MFCD00006536
:267.25 g/mol
Merck Index:14,10123
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:122.0 to 126.0 deg-C
- Specific rotation [a]20/D:+43.0 to +51.0 deg (C=1, H2O)
Specification Test Results
Related Information
Frequently Bought Together