Order Entry
Netherlands
ContactUsLinkComponent
2-(4-Aminophenoxy)naphthalene ≥98.0% (by GC, titration analysis)
2-(4-Aminophenoxy)naphthalene ≥98.0% (by GC, titration analysis)
  TCIAA2041-1G
 :  TCI
 :  
2-(4-Aminophenoxy)naphthalene ≥98.0% (by GC, titration analysis)
  TCIAA2041-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :235.29 g/mol
 :71311-83-6
 :C₁₆H₁₃NO
 :MFCD00443717