
1-(Acryloyloxy)-3-(methacryloyloxy)-2-propanol ≥80.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:1709-71-3
:
Formula:C₁₀H₁₄O₅
MDL Number:MFCD00274303
:214.22 g/mol
Flash Pt:158°C
:Stabilised with 4-methoxyphenol (MEHQ)
:GHS05
Frequently Bought Together