Order Entry
Netherlands
ContactUsLinkComponent
Fast Blue BB base ≥95.0% (by titrimetric analysis)
Irritant
Fast Blue BB base ≥95.0% (by titrimetric analysis)
Catalog # TCIAA1986-25G
Supplier:  TCI
CAS Number:  
Irritant
Fast Blue BB base ≥95.0% (by titrimetric analysis)
Catalog # TCIAA1986-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (Nonaqueous Titration):
    min. 95.0 %
  • Lambda max.:
    316.0 to 319.0nm (MeOH)
  • Absorbance (E1%1cm):
    min. 280 (20mg/L, MeOH, 316.0 to 319.0nm)

Specification Test Results

About this item

 :
 :300.36 g/mol
Melting Pt:101°C
EINECS:204-363-8
 :GHS07
CAS Number:120-00-3
Formula:C₁₇H₂₀N₂O₃
MDL Number:MFCD00009091