Order Entry
Netherlands
ContactUsLinkComponent
 
 
4-Amino-3-chlorobenzoic acid ≥97.0% (by HPLC, titration analysis)
  TCIAA1971-5G
 :  TCI
 :  
undefined
 
4-Amino-3-chlorobenzoic acid ≥97.0% (by HPLC, titration analysis)
  TCIAA1971-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 %

 

 

 :
 :171.58 g/mol
 :219-630-4
 :GHS07
 :2486-71-7
 :C₇H₆ClNO₂
 :MFCD00460423