
DL-Aminoglutethimide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:125-84-8
:
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00010122
:232.28 g/mol
Merck Index:14,00440
EINECS:204-756-4
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:150.0 to 154.0 deg-C
- Solubility in Acetone:very faint turbidity
Specification Test Results
Frequently Bought Together