4-Acetamido-2-aminobenzenesulphonic acid hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAA1944-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Water:max. 8.0 %
Specification Test Results
About this item
:
:230.24 g/mol
EINECS:201-847-0
CAS Number:88-64-2
Formula:C₈H₁₀N₂O₄S
MDL Number:MFCD00035752