O-(2-Azido-4,6-O-benzylidene-2-deoxy-ɑ-D-galactopyranosyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester ≥97.0% (by HPLC)
Catalog # TCIAA1832-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:+111.0 ~ +117.0 deg (C=1, CHCl3)
Specification Test Results
About this item
:
:672.74 g/mol
Storage Temperature:Freezer
CAS Number:195976-07-9
Formula:C₃₆H₄₀N₄O₉