4-(2-Aminoethyl)thiomorpholine-1,1-dioxide ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:89937-52-0
:
Formula:C₆H₁₄N₂O₂S
:178.25 g/mol
Boiling Pt:182°C
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:89.0 to 93.0 deg-C
Specification Test Results
Frequently Bought Together