Order Entry
Netherlands
ContactUsLinkComponent
 
1,3,5-Adamantanetriol ≥97.0% (by GC)
 :  TCI
 
1,3,5-Adamantanetriol ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Solubility in Methanol:
    almost transparency

 

 

Frequently Bought Together