Order Entry
Netherlands
ContactUsLinkComponent
1-Allyl-3,5-dimethylpyrazole ≥97.0% (by GC)
Irritant
1-Allyl-3,5-dimethylpyrazole ≥97.0% (by GC)
Catalog # TCIAA1679-5G
Supplier:  TCI
CAS Number:  
Irritant
1-Allyl-3,5-dimethylpyrazole ≥97.0% (by GC)
Catalog # TCIAA1679-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :136.20 g/mol
Boiling Pt:192°C
Flash Pt:80°C
 :GHS07
CAS Number:13369-74-9
Formula:C₈H₁₂N₂
MDL Number:MFCD00034407