
(1R,2R)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAA1657-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-38.0 to -45.0 deg (C=0.2,THF)
Specification Test Results
About this item
:
:373.41 g/mol
:GHS07
CAS Number:446044-44-6
Formula:C₂₃H₁₉NO₄