4-Acetamidocyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAA1617-25G
Supplier: TCI
CAS Number:
Specifications
- VPE:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 % (total of isomers)
Specification Test Results
About this item
:
:157.21 g/mol
Melting Pt:162°C
EINECS:245-613-6
CAS Number:23363-88-4
Formula:C₈H₁₅NO₂
MDL Number:MFCD06797038