Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2-acetamido-5-chlorobenzoate ≥98.0% (by GC)
Irritant
Methyl-2-acetamido-5-chlorobenzoate ≥98.0% (by GC)
Catalog # TCIAA1609-25G
Supplier:  TCI
CAS Number:  
Irritant
Methyl-2-acetamido-5-chlorobenzoate ≥98.0% (by GC)
Catalog # TCIAA1609-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :227.64 g/mol
 :GHS07
CAS Number:20676-54-4
Formula:C₁₀H₁₀ClNO₃
MDL Number:MFCD00144759