
Azobenzene-4,4'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:586-91-4
:
Formula:C₁₄H₁₀N₂O₄
MDL Number:MFCD00496621
:270.24 g/mol
EINECS:209-589-0
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together