
4-Amino-2-methyl-1-butanol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:44565-27-7
:
Formula:C₅H₁₃NO
MDL Number:MFCD03093615
:103.17 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together