Acetic acid (1S,2R)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenylpropyl ester ≥98.0% (by HPLC) reagent for double aldol reaction
TCIAA1535-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:119.0 to 122.0 deg-C
- Specific rotation [a]20/D:+4.0 to +6.0 deg (C=2, Chloroform)
:
:465.61 g/mol
:240423-53-4
:C₂₇H₃₁NO₄S
:MFCD02093425