Acetic acid (1R,2S)-2-[N-Benzyl-N-(mesitylenesulphonyl)amino]-1-phenylpropyl ester ≥98.0% (by HPLC) reagent for double aldol reaction
Catalog # TCIAA1534-1G
Supplier: TCI
CAS Number:
Specifications
- VPE:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:119.0 to 122.0 deg-C
- Specific rotation [a]20/D:-4.0 to -6.0 deg (C=2, Chloroform)
Specification Test Results
About this item
:
:465.61 g/mol
CAS Number:240423-74-9
Formula:C₂₇H₃₁NO₄S
MDL Number:MFCD02093424