Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Allyl butyl ether ≥97.0% (by GC)

Allyl butyl ether ≥97.0% (by GC)

25 mL
Catalog # TCIAA1481-25ML
Flammable
Irritant
Supplier:   TCI
Allyl butyl ether ≥97.0% (by GC)
Catalog # TCIAA1481-25ML
Supplier:  TCI
CAS Number:  3739-64-8
Allyl butyl ether ≥97.0% (by GC)

25 mL

CAS Number:  3739-64-8
Price
146,00 €
Log in for availability
1 * 25 mL
Product Details & Documents
Synonyms:  Allyl Butyl Ether, Butyl Allyl Ether, 1-(Allyloxy)butane
  • CAS Number:3739-64-8
  • Formula:C₇H₁₄O
  • MDL Number:MFCD00009460
  • Molecular Weight:114.19 g/mol
  • Boiling Pt:120°C
  • Flash Pt:14°C
  • EINECS:223-122-8
  • UN:3271
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specifications

  • Pk:
    25 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.