
4-Amino-6-hydroxy-2-methylpyrimidine hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:732304-23-3
Formula:C₅H₇N₃O
MDL Number:MFCD00150312
:143.14 g/mol
EINECS:212-182-0
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 98.0% (calcd.on anh.substance)
- Solubility in hot Water:very faint turbidity
- Water:max. 24.0 %
Specification Test Results
Frequently Bought Together