Order Entry
Netherlands
ContactUsLinkComponent
(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride ≥95.0% (by total nitrogen and titration analysis)
Irritant
(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride ≥95.0% (by total nitrogen and titration analysis)
Catalog # TCIAA1445-1G
Supplier:  TCI
CAS Number:  
Irritant
(S)-(-)-ɑ-Amino-γ-butyrolactone hydrochloride ≥95.0% (by total nitrogen and titration analysis)
Catalog # TCIAA1445-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 95.0 %
  • Purity (Argentmetric Titration):
    min. 95.0 %
  • Specific rotation [a]20/D:
    -25.0 to -28.0 deg (C=0.2, H2O)
  • Solubility in Water:
    very faint turbidity

Specification Test Results

About this item

 :
 :137.56 g/mol
EINECS:218-571-1
 :GHS07
CAS Number:2185-03-7
Formula:C₄H₇NO₂
MDL Number:MFCD00058172