Order Entry
Netherlands
ContactUsLinkComponent
Dimethyl-1,8-anthracenedicarboxylate ≥95.0% (by GC)
Dimethyl-1,8-anthracenedicarboxylate ≥95.0% (by GC)
Supplier:  TCI
Certificates
Dimethyl-1,8-anthracenedicarboxylate ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    102.0 to 106.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

Frequently Bought Together