Allyl-p-tolyl ether ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23431-48-3
:
Formula:C₁₀H₁₂O
MDL Number:MFCD00026098
:148.21 g/mol
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together