Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Allyl-o-tolyl ether
Allyl-o-tolyl ether ≥95.0% (by GC)
Allyl-o-tolyl ether ≥95.0% (by GC)
Allyl-o-tolyl ether ≥95.0% (by GC)
TCIAA1286-5G
:
TCI
:
Allyl-o-tolyl ether ≥95.0% (by GC)
TCIAA1286-5G
:
TCI
:
Pk:
5 g
Seq:
3173
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 95.0 %
:
:
148.21 g/mol
:
936-72-1
:
C₁₀H₁₂O
:
MFCD00026097