

Acetone 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1567-89-1
:
Formula:C₉H₁₀N₄O₄
MDL Number:MFCD00137172
:238.20 g/mol
UN:1325
ADR:4.1,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:125.0 to 128.0 deg-C
Specification Test Results
Frequently Bought Together