Order Entry
Netherlands
ContactUsLinkComponent
Acetaldehyde 2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Flammable
Irritant
Acetaldehyde 2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Flammable
Irritant
Acetaldehyde 2,4-dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    161.0 to 164.0 deg-C

Specification Test Results

Frequently Bought Together