


Diisopropyl azodicarboxylate 40% ∼1.9 mol/L in toluene
Supplier: TCI
Certificates
About this item
CAS Number:2446-83-5
:
Formula:C₈H₁₄N₂O₄
MDL Number:MFCD00008875
:202.21 g/mol
Storage Temperature:Refrigerator
EINECS:219-502-8
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid
- Concentration (Iodometric titration):40.0 to 44.0 w/w%
Specification Test Results
Frequently Bought Together