Order Entry
Netherlands
ContactUsLinkComponent
 
cis-3,3,5-Trimethylcyclohexyl acetate ≥97.0% (by GC)
  TCIAA1224-25ML
 :  TCI
 :  
cis-3,3,5-Trimethylcyclohexyl acetate ≥97.0% (by GC)
  TCIAA1224-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :184.28 g/mol
 :97°C
 :85°C
 :246-416-8
 :24691-16-5
 :C₁₁H₂₀O₂
 :MFCD00036661