Order Entry
Netherlands
ContactUsLinkComponent
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine ≥99.0% (by titrimetric analysis)
Irritant
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine ≥99.0% (by titrimetric analysis)
Catalog # TCIAA1219-1G
Supplier:  TCI
CAS Number:  
Irritant
1-Acetyl-1H-1,2,3-triazolo[4,5-b]pyridine ≥99.0% (by titrimetric analysis)
Catalog # TCIAA1219-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    112.0 to 117.0 deg-C

Specification Test Results

About this item

 :
 :162.15 g/mol
 :GHS07
CAS Number:107866-54-6
Formula:C₇H₆N₄O
MDL Number:MFCD00075383