L-Ascorbyl 2,6-dibutyrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAA1205-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:123.0 to 127.0 deg-C
- Specific rotation [a]20/D:+53.0 to +57.0 deg (C=1, EtOH)
- Solubility in EtOH:almost transparency
Specification Test Results
About this item
:
:316.31 g/mol
Storage Temperature:Refrigerator
CAS Number:4337-04-6
Formula:C₁₄H₂₀O₈
MDL Number:MFCD00144561