Disodium-4-amino-4'-nitrostilbene-2,2'-sulphonate ≥80.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6634-82-8
:
Formula:C₁₄H₁₂N₂O₈S₂
MDL Number:MFCD00143251
:444.34 g/mol
EINECS:229-631-1
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Ion exchange titration):min. 80.0 % (after deducting NaCl)
- Water:max. 10.0 %
- Sodium Chloride:max. 15.0 %
Specification Test Results
Frequently Bought Together