3-Aminopyrrolidine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:103831-11-4
:MFCD00059018
Formula:C₄H₁₀N₂
MDL Number:MFCD00060176
:159.05 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC) (CAD):min. 98.0 % (total of Ion-Pair)
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together