Order Entry
Netherlands
ContactUsLinkComponent
1,1'-(Azodicarbonyl)dipiperidine ≥98.0% (by total nitrogen basis)
Irritant
1,1'-(Azodicarbonyl)dipiperidine ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
Irritant
1,1'-(Azodicarbonyl)dipiperidine ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    134.0 to 137.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together