Atrolactic acid hydrate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:515-30-0
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00004451
:166.18 g/mol
Merck Index:14,00873
EINECS:208-196-1
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 97.0% (calcd.on anh.substance)
- Solubility in hot Water:almost transparency
- Water:max. 6.0 %
Specification Test Results
Frequently Bought Together