Order Entry
Netherlands
ContactUsLinkComponent
cis-6-Nonenyl acetate ≥92.0% (by GC)
cis-6-Nonenyl acetate ≥92.0% (by GC)
Catalog # TCIAA0921-5ML
Supplier:  TCI
CAS Number:  
cis-6-Nonenyl acetate ≥92.0% (by GC)
Catalog # TCIAA0921-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 92.0 %

Specification Test Results

About this item

 :
 :184.28 g/mol
Flash Pt:106°C
EINECS:278-397-7
CAS Number:76238-22-7
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00059409