Order Entry
Netherlands
ContactUsLinkComponent
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98.0% (by titrimetric analysis)
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0791-25G
Supplier:  TCI
CAS Number:  
undefined
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAA0791-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (Neutralization titration):
    min. 98.0% (calcd.on anh.substance)
  • Water:
    max. 12.0 %

Specification Test Results

About this item

 :
 :143.17 g/mol (anhy)
EINECS:213-722-8
CAS Number:65802-56-4
 :MFCD00150556
Formula:C₄H₅N₃OS·xH₂O
MDL Number:MFCD00052384