N-Acetyl-DL-histidine monohydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:213178-97-3
:MFCD00150969
Formula:C₈H₁₃N₃O₄
MDL Number:MFCD00065961
:215.2 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:7.5 to 10.0 %
Specification Test Results
Frequently Bought Together