
9-Aminofluorene hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5978-75-6
:
Formula:C₁₃H₁₁N
MDL Number:MFCD00012536
:217.70 g/mol
EINECS:227-778-6
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together