Order Entry
Netherlands
ContactUsLinkComponent
Amylopectin hydrate (from waxy corn) amylose-free
 :  TCI
Amylopectin hydrate (from waxy corn) amylose-free
 :  TCI
  • Appearance:
    White to Almost white powder to crystal
  • Total Nitrogen:
    max. 0.06 %
  • Ash Content:
    max. 0.1 %
  • Drying loss:
    max. 14.0 %
Frequently Bought Together