Order Entry
Netherlands
ContactUsLinkComponent
Aminopterin hydrate ≥97.0% (by HPLC, titration analysis)
Toxic
Health
Aminopterin hydrate ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Toxic
Health
Aminopterin hydrate ≥97.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Amber powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 97.0 % (calcd.on anh.substance)
  • Specific rotation [a]20/D:
    +18.0 to +22.0 deg (C=1、0.1mol/LNaOH sol.) (calcd.on anh.substance)
  • Water:
    3.0 to 10.0 %

Specification Test Results

Related Information

Frequently Bought Together