
6-Amino-4-hydroxy-2-naphthalenesulphonic acid ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAA0371-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 % (calcd.on anh.substance)
- Water:max. 10.0 %
Specification Test Results
About this item
:
:239.25 g/mol
EINECS:202-000-8
:GHS07
CAS Number:90-51-7
Formula:C₁₀H₉NO₄S
MDL Number:MFCD00003973