Order Entry
Netherlands
ContactUsLinkComponent
Disodium anthraquinone-2,6-disulphonate ≥98.0% (by HPLC, titration analysis)
Disodium anthraquinone-2,6-disulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0308-25G
Supplier:  TCI
CAS Number:  
Disodium anthraquinone-2,6-disulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0308-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 % (calcd.on dried substance)
  • Solubility in hot Water:
    almost transparency

Specification Test Results

About this item

 :
 :412.29 g/mol
EINECS:212-719-9
CAS Number:853-68-9
Formula:C₁₄H₈O₈S₂
MDL Number:MFCD00001230