N-Acetyl-L-tryptophan ethyl ester ≥98.0% (by HPLC, total nitrogen)
Catalogus # TCIAA0122-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:106.0 to 111.0 deg-C
- Specific rotation [a]20/D:+5.0 to +8.0 deg (C=2, EtOH)
Specification Test Results
About this item
:
:274.32 g/mol
EINECS:219-190-3
CAS Number:2382-80-1
Formula:C₁₅H₁₈N₂O₃
MDL Number:MFCD00005643