(S)-(-)-1-Phenylethyl isocyanate, Supelco®
Catalog # 77970-1ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:147.18 g/mol
Boiling Pt:55 °C (1013 hPa)
Density:1.05 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:238-698-6
CAS Number:14649-03-7
Formula:C₉H₉NO
MDL Number:MFCD00002036