L(+)-Lysine dihydrochloride 99%
:657-26-1
:
:C₆H₁₆Cl₂N₂O₂
:MFCD00066389
:219.11 g/mol
:202 °C
:211-518-3
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 3 h)
- Specific optical rotation:+14° to +18° (20°C, 589 nm) (c=2, H2O)
- Titration with HClO4:98.5 to 101.5 % (on dried substance)
Frequently Bought Together