Order Entry
Netherlands
ContactUsLinkComponent
Tin(IV) chloride 1 mol/l in dichloromethane, AcroSeal®
Irritant
Health
Corrosive
Tin(IV) chloride 1 mol/l in dichloromethane, AcroSeal®
Certificates
undefined
Irritant
Health
Corrosive
Tin(IV) chloride 1 mol/l in dichloromethane, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Solution
  • Titration Argentometric:
    0.95 to 1.10 M